Learn More
Leica Biosystems Activated charcoal filter set for Spectra CV and Spectra ST microscopes
Supplier:  Leica Biosystems 14051253772
Encompass
Activated charcoal filter set designed as a replacement air filtration component for Spectra CV and Spectra ST laboratory microscopes. The activated charcoal media adsorbs odors and airborne contaminants from instrument airflow, helping maintain a clean internal environment and protect sample integrity. Intended for routine maintenance and scheduled filter replacement.
  • Pack of two replacement activated charcoal filters.
  • Adsorbs odors and airborne contaminants from instrument airflow.
  • Helps maintain a clean internal atmosphere and protect sample integrity.
  • Suitable for routine maintenance and scheduled filter replacement.
  • New OEM condition as supplied by authorized distributors.
Catalog No. NC1744746