Learn More
Sigma Aldrich Fine Chemicals Biosciences Activated Charcoal Norit(R) Norit GAC 1240W, from coal, for potable water processing, steam activated, granular | 7440-44-0 | MFCD00133992 | 1KG
Catalog No. 501879597
Encompass
Activated Charcoal Norit(R) Norit GAC 1240W, from coal, for potable water processing, steam activated, granular | Mol Wt: 12.01 | 7440-44-0 | MFCD00133992 | 1KG
Catalog No. 50-187-9597 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 377711KG
Activated Charcoal Norit(R) Norit GAC 1240W, from coal, for potable water processing, steam activated, granular | Mol Wt: 12.01 | 7440-44-0 | MFCD00133992 | 1KG