Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activated charcoal puriss. p.a., powder, MilliporeSigma™ Supelco™

Catalog No. 111015411 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5411 250 g
11-101-5410 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5411 Supplier MilliporeSigma Supelco Supplier No. 31616250G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Activated charcoal is an FDA approved adsorbent and generally finds application in being used during the treatment of swallowed poison.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00133992
Synonym Charcoal activated

Specifications

Melting Point 3550°C
CAS 7440-44-0
pH 6 to 9
Ignition Point 842°F
Linear Formula C
Quantity 250 g
Vapor Pressure <0.1 mmHg (20°C)
Formula Weight 12.01
Grade Puriss. p.a.
Physical Form Powder
Chemical Name or Material GC Accessories & Adsorbents
Show More Show Less
RTECSNumber FF5250100
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.