Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Activated Penetratin™

Catalog No. MP1PENA0500 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1PENA0500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1PENA0500 Supplier MP Biomedicals™ Supplier No. 11PENA0500

missing translation for 'validMainframeMsg'

Successfully used to internalize covalently attached peptides, oligonucleotides and siRNA.

Applications: Activated PenetratinTM 1 Peptide is a novel tool for peptide and siRNA delivery. It has been successfully used to internalize covalently attached peptides, oligonucleotides, and siRNA. It is sold ready for coupling to peptides or oligonucleotides containing thiol (-SH) functions.
Product Overview: PenetratinTM 1 is a patented 16-amino acid peptide corresponding to the third helix of the homeodomain of antennapedeia protein. This peptide is able to translocate across biological membranes by an energy-independent mechanism (1-3) and has been used successfully to internalize covalently attached peptides (4), oligonucleotides (5) and siRNA (6). Activated PenetratinTM 1 is sold ready for coupling to peptides or oligos containing thiol (-SH) functions. Biotinylated Activated PenetratinTM 1 possesses a biotin group at the C-terminal end allowing the detection of the internalized molecule with an appropriate avidin or streptavidin reagent.
Product Description: The Active PenetratinTM 1 Peptide Transfection reagent features a patented 16 amino acid peptide corresponding to the third helix of the homeodomain of the Antennapedia protein (pAntp) from Drosophila melanogaster.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Stable for two years when frozen (<-15 °C).
Formula Weight 2588
For Use With (Application) Transfection
Product Type Reagents
Quantity 500 μg
Synonym Transfection Reagent
Form Lyophilized Powder

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.