Learn More
Medchemexpress LLC HY-P1918 5mg Medchemexpress, Activated Protein C (390-404), human CAS:146340-20-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP19185MG
Encompass_Preferred
Medchemexpress, HY-P1918 5mg Activated Protein C (390-404), human CAS:146340-20-7 Activated Protein C (390-404), human is a peptide of the activated protein C (a vitamin K-dependent serine protease), potently inhibits APC anticoagulant activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3078