Learn More
Sigma Aldrich Activation Kit For Multiplex Microspheres
Catalog No. ALMPAKMM Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
ALMPAKMM 5 mL
1 options
Catalog No. ALMPAKMM Supplier Sigma Aldrich Supplier No. ALMPAKMM10RXN
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
AnteoTech identified the following problems that researchers are having with their multiplex assays:Binding difficult antibodies and antigens to microspheresComplex protocols for inexperienced usersLengthy microsphere and protein preparation times prior to protein couplingReproducibility between batches of protein coupled microspheresLimited storage of ready-to-couple microspheresFrom these findings, they developed the AnteoBind(TM) Activation Kit For Multiplex Microspheres that is for use with the Luminex xMAP™ system and contains the patented AnteoBind Activation Reagent. With this Activation Kit, you can couple multiple antibodies without extra optimization for each, saving assay development time and reducing the barriers of assay transfer between diverse platforms.AnteoBind is water based and replaces the dry chemicals you would use with the EDC method. Using AnteoBind instead of NHS/EDC chemistry can save up to 3-4 hours of lab time. Experiments have shown increased stability and functionality for antibodies and a decreased background. AnteoBind improves reproducibility between experiments by providing uniformity between surfaces. You can store your AnteoBind activated microspheres for up to 1 year.POSTER: A Novel User Friendly Chemistry to Couple Antibodies and Challenging Proteins to Luminex MagPlex MicrospheresActivation and coupling of Luminex Microspheres for Multiplex ImmunoassaysxMAP is a registered trademark of Luminex Corp
Specifications
CAS 1622426-20-3
Quantity 5 mL