Learn More
Signosis Inc ER Stress (UPR) TF Activation Profiling Plate Array
Catalog No. NC1334869
Encompass
The Unfolded Protein Response (UPR) is a conserved and essential stress response that cells activate to combat endoplasmic reticulum (ER) stress, commonly caused by the accumulation of misfolded proteins or failing protein quality control. ER stress is a well-characterized feature of several diseases, such as diabetes, Alzheimer’s, Parkinson’s, Huntingon’s, and prion diseases. Early cellular response to ER stress includes the transcriptional upregulation of chaperone proteins, which is mediated by a large number of transcription factors (TFs).
Catalog No. NC1334869 Supplier Signosis Inc Supplier No. FA1006
The Unfolded Protein Response (UPR) is a conserved and essential stress response that cells activate to combat endoplasmic reticulum (ER) stress, commonly caused by the accumulation of misfolded proteins or failing protein quality control. ER stress is a well-characterized feature of several diseases, such as diabetes, Alzheimer’s, Parkinson’s, Huntingon’s, and prion diseases. Early cellular response to ER stress includes the transcriptional upregulation of chaperone proteins, which is mediated by a large number of transcription factors (TFs).