Learn More
IPHASE PHARMA SERVICES LLC Activation System,Rat Liver S9 Mix,10mL
Catalog No. NC3966966
Encompass_Preferred
The induced rat liver S9 activation system was prepared from the livers of SD rats induced by the combination of sodium phenobarbital and β-naphthoflavone, containing cofactors required for metabolism. This system contains induced SD rat liver S9, Magnesium-potassium solution, D-Glucose 6-phosphate solution, Coenzyme II solution, and PBS. The protein concentration for this system is 3.5 mg/mL.
Catalog No. NC3966966 Supplier IPHASE PHARMA SERVICES LLC Supplier No. 0271031
The induced rat liver S9 activation system was prepared from the livers of SD rats induced by the combination of sodium phenobarbital and β-naphthoflavone, containing cofactors required for metabolism. This system contains induced SD rat liver S9, Magnesium-potassium solution, D-Glucose 6-phosphate solution, Coenzyme II solution, and PBS. The protein concentration for this system is 3.5 mg/mL.