Learn More
Roche Diagnostics System reagent for the Cedex Bio and the Cedex Bio HT Analyzers
Catalog No. NC1140070 Shop All Roche Diagnostics Products
Encompass
System reagent for the Cedex Bio and the Cedex Bio HT Analyzers. Used for preparation of the pipetting probes in the daily initialization procedure of the instrument, and for activation of the membranes in the ISE module.
Catalog No. NC1140070 Supplier Roche Diagnostics Supplier No. 07766700001
System reagent for the Cedex Bio and the Cedex Bio HT Analyzers. Used for preparation of the pipetting probes in the daily initialization procedure of the instrument, and for activation of the membranes in the ISE module.