Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Burdick & Jackson™ Activator Reagent, 0.5M ETT in Acetonitrile

Catalog No. p-7161164 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2 x 1 L
4 x 2.5 L
Packaging Type:
GLASS BOTTLE, GL45
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
60-048-335 2 x 1 L Glass Bottle
60-048-336 4 x 2.5 L GLASS BOTTLE, GL45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-335 Supplier Honeywell Burdick & Jackson™ Supplier No. BR7251
Only null left
Add to Cart
Add to Cart
Request a Quote

BioSyn™, for DNA/RNA synthesis

Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis

  • Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-05-8
Density 0.809 g/cm3
Packaging Type Glass Bottle
Quantity 2 x 1 L
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.