Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activator Reagent, 0.5M ETT in Acetonitrile, MilliporeSigma™

Catalog No. MBI01582505 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MBI01582505 2.5 L Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MBI01582505 Supplier MilliporeSigma Supplier No. BI01582505
  • Involved in formulations for oligonucleotide synthesis
  • Suitable for all kind of biosynthesis processes such as DNA synthesis
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ETT in Acetonitrile
Color Colorless
Flash Point 5.6°C
Packaging Plastic bottle
Quantity 2.5 L
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
Formula Weight Mixture of organic compounds
Concentration or Composition (by Analyte or Components) 0.48 to 0.52M
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.