Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activator Reagent,Solstice

Catalog No. 60090110 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
20 L
4 x 2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-110 20 L
60-090-020 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-090-110 Supplier Solstice Supplier No. BR72120
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.25M ETT in Acetonitrile

Solstice BioSyn™ reagents and solvents for DNA/RNA synthesis

  • BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY

Specifications

CAS 89797-68-2, 75-05-8
CAS Max % 4.00%, 96.00%
Quantity 20 L
Flash Point 7°C
Packaging Returnable NOWPAK™ II
Physical Form Liquid
Purity Grade Notes Activator reagents
Synonym 5-(Ethylthio)-1H-Tetrazole, 5-(Ethylthio)Tetrazole Solution
Boiling Point 82°C.The physical data is that of the main component.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.