Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Activator Solution 0.25M 5-(Ethylthio)-1H-Tetrazole in Acetonitrile, MilliporeSigma™

Catalog No. 50947746 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-746 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-746 Supplier MilliporeSigma Supplier No. BI01521005
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89797-68-2
Molecular Formula C3H6N4S
Molecular Weight (g/mol) 130.17
InChI Key GONFBOIJNUKKST-UHFFFAOYSA-N
IUPAC Name 5-(ethylsulfanyl)-2H-1,2,3,4-tetrazole
SMILES CCSC1=NNN=N1

Specifications

Boiling Point 81.6°C (1013 hPa) (Acetonitrile)
Percent Purity ≥99.9%
Packaging Glass Bottle
Vapor Pressure 97 hPa (20°C)
Quantity 1 L
Flash Point 7.2°C
Recommended Storage Store below 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.