Learn More
Mitegen Active 4L Dewar Drying Rack for LN2 Dewars
SDP
Catalog No. 5000494988
Encompass
The Active 4L Dewar Drying Rack offers a fast and reliable way to dry your LN2 dewars in just 35 minutes. This rack features a heated, filtered airflow system that automatically senses a dewar and initiates the drying cycle. The integrated LED display provides clear cycle status, ensuring your dewars are completely dry and contaminant-free for your next experiment. The rack enhances lab productivity by eliminating downtime associated with air-drying dewars. Simply place your cleaned 4L dewar on the rack with the drying tube inserted, and the process begins automatically. A short cool-down period is included, bringing the dewar to room temperature. The Active 4L Dewar Drying Rack is compatible with both MVE and ICBiomedical 4L dewars. Once the drying process is complete, the dewar can be transferred to the Passive Drying and Storage Rack SKU M-CEM-NS-PDR-LD4-1 for secure storage.
Catalog No. 50-004-94988 Supplier Mitegen Supplier No. MCEMNSADRLD41
The Active 4L Dewar Drying Rack offers a fast and reliable way to dry your LN2 dewars in just 35 minutes. This rack features a heated, filtered airflow system that automatically senses a dewar and initiates the drying cycle. The integrated LED display provides clear cycle status, ensuring your dewars are completely dry and contaminant-free for your next experiment. The rack enhances lab productivity by eliminating downtime associated with air-drying dewars. Simply place your cleaned 4L dewar on the rack with the drying tube inserted, and the process begins automatically. A short cool-down period is included, bringing the dewar to room temperature. The Active 4L Dewar Drying Rack is compatible with both MVE and ICBiomedical 4L dewars. Once the drying process is complete, the dewar can be transferred to the Passive Drying and Storage Rack SKU M-CEM-NS-PDR-LD4-1 for secure storage.