Learn More
Crystal Chem Inc Active GIP ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 81509
Encompass
The Active GIP ELISA is a highly sensitive assay that is used to quantify levels of human GIP active in plasma and cell culture. Highlights: Kits use a sample volume of only 50 µL. High Sensitivity of 0.78 pM. Dynamic Range of 0.78 - 50 pM. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7895