Learn More
Abcam Recombinant human EGF protein (Animal Free), 500UG
Supplier:  Abcam AB9697500UG
Encompass_Preferred
Recombinant human EGF protein (Animal Free) is a Human Full Length protein, in the 971 to 1023 aa range, expressed in Escherichia coli, with >=98% purity, < 0.01 EU/µg endotoxin level and suitable for SDS-PAGE, WB, HPLC, CellAct.
Protein/ Peptide/ Enzyme Synonyms: Pro-epidermal growth factor, EGF
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0735255