Learn More
Sciencix Sciencix Active Inlet Valve Cartridge, 600 Bar Pressure Rating, Compatible With Agilent HPLC Systems, Each
SDP
Catalog No. NC3590223
Encompass_Preferred
The SCIENCIX Active Inlet Valve Cartridge is designed for high-performance HPLC systems, ensuring reliable solvent delivery at pressures up to 600 bar. Made with durable wetted materials like sapphire and stainless steel, it maintains chemical resistance and system integrity. Its compatibility with Agilent HPLC systems like the 1260 and G1310 series offers users a reliable replacement for OEM parts, differentiating it with a lifetime warranty against manufacturing defects.
Catalog No. NC3590223 Supplier Sciencix Supplier No. 114145
The SCIENCIX Active Inlet Valve Cartridge is designed for high-performance HPLC systems, ensuring reliable solvent delivery at pressures up to 600 bar. Made with durable wetted materials like sapphire and stainless steel, it maintains chemical resistance and system integrity. Its compatibility with Agilent HPLC systems like the 1260 and G1310 series offers users a reliable replacement for OEM parts, differentiating it with a lifetime warranty against manufacturing defects.