Learn More
Agilent Technologies ACTIVE INLET VALVE WITHOUT CA
Catalog No. NC2163689
Encompass
  • Active inlet valve designed for HPLC systems
  • No cartridge included
  • Compatible with Agilent 1100, 1200, 1200 RRLC, 1260 Infinity systems
  • Regulates solvent flow effectively
  • Prevents back-flushing during operation
  • Eliminates vapor formation with volatile solvents
  • Maintains optimal pressure and flow for precise analysis
  • Ideal for high-throughput environments
  • Supports analytical applications in various laboratories
  • Pressure rating compatible up to 600 bar
This item is not returnable. View return policy
Catalog No. NC2163689 Supplier Agilent Technologies Supplier No. G131260025
  • Active inlet valve designed for HPLC systems
  • No cartridge included
  • Compatible with Agilent 1100, 1200, 1200 RRLC, 1260 Infinity systems
  • Regulates solvent flow effectively
  • Prevents back-flushing during operation
  • Eliminates vapor formation with volatile solvents
  • Maintains optimal pressure and flow for precise analysis
  • Ideal for high-throughput environments
  • Supports analytical applications in various laboratories
  • Pressure rating compatible up to 600 bar