Learn More
Active Motif Chromeo P503 Fluorogenic Dye, Amine-Reactive, Bright Color Change, 1 mg
SDP
Catalog No. NC1875285
Encompass
Chromeo P503 is a fluorogenic dye that labels proteins and peptides by binding to primary amine groups, providing clear detection with its distinct blue to red color change. Its quantum yield dramatically increases upon conjugation, enabling effective biomolecule labeling suitable for diverse applications like gel electrophoresis and chromatography. This dye distinguishes itself with a significant shortwave spectral shift, minimizing background effects for reproducible results.
Catalog No. NC1875285 Supplier Active Motif Supplier No. 15106
Chromeo P503 is a fluorogenic dye that labels proteins and peptides by binding to primary amine groups, providing clear detection with its distinct blue to red color change. Its quantum yield dramatically increases upon conjugation, enabling effective biomolecule labeling suitable for diverse applications like gel electrophoresis and chromatography. This dye distinguishes itself with a significant shortwave spectral shift, minimizing background effects for reproducible results.