Learn More
Waters Corp Active Preheater (APH), MP35N, 0.005 ID, 12.5 in. Length
Catalog No. 502391834 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement active preheater (APH) is made of MP35N material, with a 0.005 ID and a length of 12.5 inches. It is utilized within ACQUITY Arc and ACQUITY Arc Bio CH-A and CM-A with 2 and 4 columns.
  • Made from MP35N material
  • Features a 0.005 ID
  • Measures 12.5 in. long
  • Supplied as a single unit
  • Mechanical type component
  • Compatible with UPLC systems
  • Utilized within ACQUITY Arc and ACQUITY Arc Bio CH-A and CM-A with 2 and 4 columns
Catalog No. 50-239-1834 Supplier Waters Corp Supplier No. 205001803
This replacement active preheater (APH) is made of MP35N material, with a 0.005 ID and a length of 12.5 inches. It is utilized within ACQUITY Arc and ACQUITY Arc Bio CH-A and CM-A with 2 and 4 columns.
  • Made from MP35N material
  • Features a 0.005 ID
  • Measures 12.5 in. long
  • Supplied as a single unit
  • Mechanical type component
  • Compatible with UPLC systems
  • Utilized within ACQUITY Arc and ACQUITY Arc Bio CH-A and CM-A with 2 and 4 columns