Learn More
Waters Corp Active PreHeater, MP35N, 0.004 in ID x 18.5 in
Catalog No. 502392950 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Active PreHeater (APH) is used in H-Class PLUS Bio systems with multiple CM-A's. This component is crucial for maintaining optimal system performance.
  • Material: MP35N
  • Dimensions: 0.004 in ID x 18.5 in
  • Compatible with H-Class PLUS Bio systems.
  • Includes a gasket for installation.
  • Designed as a mechanical spare part.
  • Suitable for UPLC system types.
  • Replacement is recommended as required.
Catalog No. 50-239-2950 Supplier Waters Corp Supplier No. 205001755
The Active PreHeater (APH) is used in H-Class PLUS Bio systems with multiple CM-A's. This component is crucial for maintaining optimal system performance.
  • Material: MP35N
  • Dimensions: 0.004 in ID x 18.5 in
  • Compatible with H-Class PLUS Bio systems.
  • Includes a gasket for installation.
  • Designed as a mechanical spare part.
  • Suitable for UPLC system types.
  • Replacement is recommended as required.