Learn More
Waters Corp Waters Active Pre-Heater, Stainless Steel, .003 in ID x 12.5 in
Catalog No. 502039278 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This is a replacement active pre-heater designed for use in I-Class PLUS Bio systems with CM-A. It is constructed from stainless steel and includes a gasket.
  • Stainless steel construction.
  • Dimensions of .003 in. ID × 12.5 in.
  • Includes a gasket.
  • Compatible with I-Class PLUS Bio systems with CM-A.
  • Functions as system tubing.
  • A component for the I-Class system's column heater.
  • Designed for as-needed replacement.
Catalog No. 50-203-9278 Supplier Waters Corp Supplier No. 205001757
This is a replacement active pre-heater designed for use in I-Class PLUS Bio systems with CM-A. It is constructed from stainless steel and includes a gasket.
  • Stainless steel construction.
  • Dimensions of .003 in. ID × 12.5 in.
  • Includes a gasket.
  • Compatible with I-Class PLUS Bio systems with CM-A.
  • Functions as system tubing.
  • A component for the I-Class system's column heater.
  • Designed for as-needed replacement.