Learn More
Waters Corp Waters Corporation PreHeater, Stainless Steel, 0.007 in. ID, 36.5 in. Length
Catalog No. 502391572 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit is designed for use with the ACQUITY UPC2 system. It features durable stainless steel construction and precise dimensions, making it suitable for specific applications requiring a 0.007 in. internal diameter and a 36.5 in. length.
  • Durable stainless steel construction
  • Designed for ACQUITY UPC2 system
  • Internal diameter of 0.007 in.
  • Length of 36.5 in.
Catalog No. 50-239-1572 Supplier Waters Corp Supplier No. 205001004
This kit is designed for use with the ACQUITY UPC2 system. It features durable stainless steel construction and precise dimensions, making it suitable for specific applications requiring a 0.007 in. internal diameter and a 36.5 in. length.
  • Durable stainless steel construction
  • Designed for ACQUITY UPC2 system
  • Internal diameter of 0.007 in.
  • Length of 36.5 in.