Learn More
Waters Corp Waters Corporation Active Preheater, Stainless Steel, 12.5 in., ACQUITY UPLC
Catalog No. 50791696 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement active preheater is constructed from stainless steel and measures 12.5 in. long. It is designed for use within ACQUITY CM-A, CH-A, and ACQUITY APC CM-S systems.
  • Units per package: 1
  • Type: Tubing
  • System type: UPLC
  • Instrument: H-Class system
  • Replacement schedule: As required
  • Component: Column manager, column heater
Catalog No. 50-791-696 Supplier Waters Corp Supplier No. 205000730
This replacement active preheater is constructed from stainless steel and measures 12.5 in. long. It is designed for use within ACQUITY CM-A, CH-A, and ACQUITY APC CM-S systems.
  • Units per package: 1
  • Type: Tubing
  • System type: UPLC
  • Instrument: H-Class system
  • Replacement schedule: As required
  • Component: Column manager, column heater