Learn More
Creative Biomart Active Recombinant Human TNFSF8, Fc tagged, 100ug
Catalog No. NC3356425
Encompass
Recombinant Human TNFSF8 extracellular domain (Gln 63-Asp 234) (NP_001235.1), fused with the Fc region of human IgG1 at the N-terminus, was produced in Human Cell.
Catalog No. NC3356425 Supplier Creative Biomart Supplier No. TNFSF860H100UG
Recombinant Human TNFSF8 extracellular domain (Gln 63-Asp 234) (NP_001235.1), fused with the Fc region of human IgG1 at the N-terminus, was produced in Human Cell.