Learn More
Cell Signaling Technology Active Rho Detection Kit, RhoA, Human/Mouse, 1 Kit
SDP
Catalog No. 502730421
Encompass
The Active Rho Detection Kit provides all necessary reagents for measuring the activation of Rho GTPase in cells. The system utilizes a GST-Rhotekin-RBD fusion protein to selectively bind the activated, GTP-bound form of Rho. This complex is then isolated via immunoprecipitation with glutathione resin, allowing for the subsequent determination of Rho activation levels through western blot analysis with a specific Rho rabbit antibody.
  • Detects endogenous levels of GTP-bound (active) Rho.
  • Includes GST-Rhotekin-RBD fusion protein, glutathione resin, Rho rabbit antibody, and essential buffers for 30 immunoprecipitations.
  • Validated for use with human and mouse samples.
  • Suitable for functional analysis applications.
Catalog No. 50-273-0421 Supplier Cell Signaling Technology Supplier No. 8820S
The Active Rho Detection Kit provides all necessary reagents for measuring the activation of Rho GTPase in cells. The system utilizes a GST-Rhotekin-RBD fusion protein to selectively bind the activated, GTP-bound form of Rho. This complex is then isolated via immunoprecipitation with glutathione resin, allowing for the subsequent determination of Rho activation levels through western blot analysis with a specific Rho rabbit antibody.
  • Detects endogenous levels of GTP-bound (active) Rho.
  • Includes GST-Rhotekin-RBD fusion protein, glutathione resin, Rho rabbit antibody, and essential buffers for 30 immunoprecipitations.
  • Validated for use with human and mouse samples.
  • Suitable for functional analysis applications.