Learn More
Abcam Recombinant S. japonicum GST protein, 100UG
Supplier:  Abcam AB168026100UG
Encompass_Preferred
Recombinant S. japonicum GST protein is a Schistosoma japonicum Full Length protein, in the 1 to 218 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: Glutathione S-transferase class-mu 26 kDa isozyme, GST 26, Sj26 antigen, SjGST
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1024526