Learn More
ACROBiosystems ACROBiosystems ActiveMax Human VEGF165 Protein, Tag Free (MALS verified)
Catalog No. 502106780
Encompass_Preferred
Recombinant Protein;1MG;ActiveMax Human VEGF165, Tag Free (MALS verified) (VE5-H4210) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession # P15692-4).;This protein carries no "tag". The protein has a calculated MW of 19.2 kDa. As a result of glycosylation, the protein migrates as 24 kDa (monomer) under reducing (R) condition, and 43-50 kDa (homodimer) under non-reducing (NR) condition (SDS-PAGE).;Immobilized ActiveMax Human VEGF165, Tag Free (MALS verified) (Cat. No. VE5-H4210) at 0.01 μg/mL (100 μL/well) can bind Human VEGF R1, His Tag (Cat. No. VE1-H5220) with a linear range of 4-31 ng/mL (QC tested).;Immobilized ActiveMax Human VEGF165, Tag Free (MALS verified) (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Biotinylated Human VEGF R2, Avitag,His Tag (Cat. No. KDR-H82E5) with a linear range of 10-156 ng/mL (Routinely tested).;Avastin (Bevacizumab) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind ActiveMax Human VEG
Catalog No. 50-210-6780 Supplier ACROBiosystems Supplier No. VE5H42101MG
Recombinant Protein;1MG;ActiveMax Human VEGF165, Tag Free (MALS verified) (VE5-H4210) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 191 (Accession # P15692-4).;This protein carries no "tag". The protein has a calculated MW of 19.2 kDa. As a result of glycosylation, the protein migrates as 24 kDa (monomer) under reducing (R) condition, and 43-50 kDa (homodimer) under non-reducing (NR) condition (SDS-PAGE).;Immobilized ActiveMax Human VEGF165, Tag Free (MALS verified) (Cat. No. VE5-H4210) at 0.01 μg/mL (100 μL/well) can bind Human VEGF R1, His Tag (Cat. No. VE1-H5220) with a linear range of 4-31 ng/mL (QC tested).;Immobilized ActiveMax Human VEGF165, Tag Free (MALS verified) (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Biotinylated Human VEGF R2, Avitag,His Tag (Cat. No. KDR-H82E5) with a linear range of 10-156 ng/mL (Routinely tested).;Avastin (Bevacizumab) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind ActiveMax Human VEG