Learn More
ACROBiosystems Recombinant Protein;ActiveMax Recombinant Human GM-CSF;HEK293;20UG;GMF,GM-CSF,GMCSF,CSF2,CSF-2,MGC131935,MGC138897
Catalog No. 501138346
Encompass_Preferred
Recombinant Protein;ActiveMax Recombinant Human GM-CSF;20UG;Human GM-CSF, Tag Free (GMF-H4214) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - Glu 144 (Accession # NP_000749.2).;This protein carries no "tag". The protein has a calculated MW of 14.5 kDa. The protein migrates as 16-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;The bio-activity was determined by dose-dependent stimulation of the proliferation of TF-1 cells.The ED50 < 0.1 ng/ml, corresponding to a specific activity of >1x107 Unit/mg.
Catalog No. 50-113-8346 Supplier ACROBiosystems Supplier No. GMFH421420UG
Recombinant Protein;ActiveMax Recombinant Human GM-CSF;20UG;Human GM-CSF, Tag Free (GMF-H4214) is expressed from human 293 cells (HEK293). It contains AA Ala 18 - Glu 144 (Accession # NP_000749.2).;This protein carries no "tag". The protein has a calculated MW of 14.5 kDa. The protein migrates as 16-26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;The bio-activity was determined by dose-dependent stimulation of the proliferation of TF-1 cells.The ED50 < 0.1 ng/ml, corresponding to a specific activity of >1x107 Unit/mg.