Learn More
ACROBiosystems Recombinant Protein;ActiveMax Recombinant Human VEGF110 (HPLC-verified);HEK293;20UG;VE0,RP1-261G23.1,MGC70609,MVCD1,MVCD-1,VEGF 165,VEGF-A,VEGFA,VPF,VEGF165,VEGF-165,VEGF121,VEGF120,VEGF186,VEGF164,VEGF110
Supplier:  ACROBiosystems VE0H521220UG
Encompass_Preferred
Recombinant Protein;ActiveMax Recombinant Human VEGF110 (HPLC-verified);20UG;Human VEGF110, Tag Free (VE0-H5212) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 136 (Accession # NP_001165097).;This protein carries no "tag". The protein has a calculated MW of 12.6 kDa. As a result of glycosylation, the protein migrates as 18-20 kDa under reducing (R) condition, and 40-45 kDa (homodimer) under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human VEGF110, Tag Free (Cat. No. VE0-H5212) at 0.05 g/mL (100 L/well) can bind Human VEGF R1, His Tag (Cat. No. VE1-H5220) with a linear range of 4-63 ng/mL (QC tested).
Catalog No. 50-113-8208