Learn More
Enzo Life Sciences Activin-A (human), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX2018090010
Encompass_Preferred
Alternative name: Inhibin beta-1, FRP, FSH-releasing protein. Endotoxin: ≤1 EU/μg protein measured by kinetic LAL analysis. Purity: ≥95% (HPLC, Reducing and Non-reducing SDS-PAGE, UV spectroscopy at 280 nm). MW: ~26.2kDa. Appearance: White lyophilized (freeze-dried) powder. Formulation: Lyophilized from a sterile filtered solution containing 0.1% TFA. Source: Produced in E. coli. Non-glycosylated homodimer, containing two 117 amino acid chains. Long Term Storage: -20°C. Use/Stability: It is recommended that a carrier protein (0.1% HSA or BSA) is added for long term storage. Handling: Centrifuge the vial before opening the cap. After reconstitution, prepare aliquots and store at -20°C.
Catalog No. 50-200-8013