Learn More
Medchemexpress LLC Activin A Human/Mou 50ug
SDP
Catalog No. 5000180233
Encompass_Preferred
Activin A a multifunctional cytokine is a member of TGF- superfamily Activin A first binds to the type II activin receptors (ActIIRA or ActRIIB) on the member surface and then recruits and phosphorylates type I activin receptors (ActRI) Activin A primarily signal through SMAD2/3 proteins to regulate a variety of functions including inflammation fibrosis and tumorigenesis Activin A Protein Mouse (HEK293 His) is produced in HEK293 cells with a C-Terminal His-tag
Catalog No. 50-001-80233 Supplier Medchemexpress LLC Supplier No. HYP7324150UG
Activin A a multifunctional cytokine is a member of TGF- superfamily Activin A first binds to the type II activin receptors (ActIIRA or ActRIIB) on the member surface and then recruits and phosphorylates type I activin receptors (ActRI) Activin A primarily signal through SMAD2/3 proteins to regulate a variety of functions including inflammation fibrosis and tumorigenesis Activin A Protein Mouse (HEK293 His) is produced in HEK293 cells with a C-Terminal His-tag