Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci Activin A Recombinant Protein
SDP

Catalog No. 16838502
Change view
Click to view available options
Quantity:
0.002 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-838-502 0.002 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-838-502 Supplier ProSci Supplier No. 402120002
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Activin A is a TGF-β family member that exhibits a wide range of biological activities including regulation of cellular proliferation and differentiation, and promotion of neuronal survival. Elevated levels of Activin A in human colorectal tumors and in post-menopausal woman have been implicated in colorectal and breast cancers, respectively. The biological activities of Activin A can be neutralized by inhibins and by the diffusible TGF-β antagonist, Follistatin. Human Activin A is a 26.0 kDa disulfide-linked homodimer of two β A chains, each containing 116 amino acid residues.

Specifications

Accession Number NP_001003652.1
Gene ID (Entrez) 4087
Quantity 0.002 mg
Source (BTI-Tn-5B1-4) High-5 Insect cells
Common Name SMAD family member 2
Gene Symbol SMAD2
Biological Activity The ED50 was determined by its ability to inhibit the proliferation of murine MPC - 11 cells is ≤ 2 ng/mL, corresponding to a specific activity of ≥ 5 x 105 units/mg.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence GLECDGKVNI CCKKQFFVSF KDIGWNDWII APSGYHANYC EGECPSHIAG TSGSSLSFHS TVINHYRMRG HSPFANLKSC CVPTKLRPMS MLYYDDGQNI IKKDIQNMIV EECGCS
Content And Storage The lyophilized Activin A recombinant protein is stable for at least 2 years from date of receipt at -20°C. Reconstituted Activin A is stable for at least 3 months when stored in working aliquots with a carrier protein at -20°C. As with any protein, exposing Activin A recombinant protein to repeated freeze / thaw cycles is not recommended. When working with proteins care should be taken to keep recombinant protein (at a cool and stable temperature.)
Purity or Quality Grade >95% by SDS-PAGE gel and HPLC analyses. Endotoxin level is less than 0.1 ng / μg (1 EU/μg).
Synonym JV18, MADH2, MADR2, JV18-1, hMAD-2, hSMAD2, Mothers against decapentaplegic homolog 2, MAD homolog 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.