Learn More
Vector Laboratories ACTIVIN QUANT ELISA KIT
SDP
Supplier:  Vector Laboratories LSF6867MSS121
Encompass
Human INHBE Activin (Sandwich ELISA) ELISA Kit.S-F6867 is a 96-well enzyme-linked immunosorbent assay (ELISA) for the Quantitative detection of Human INHBE / Activin in samples of Cell Culture Supernatants, Cell Lysates, Plasma, Serum and Tissue Homogenates.
Catalog No. NC2272282