Learn More
Creative Biomart Recombinant Hu-rhEGF-rP64k/Mont Therapeutic Vaccine
Catalog No. 502084840
Encompass_Preferred
Hu-rhEGF-rP64k/Mont is a therapeutic vaccine composed by an antigen (recombinant human EGF (rhEGF) chemically conjugated to recombinant P64K (rP64K)) and an adjuvant (Montanide ISA51VG) being investigated for the treatment of advanced non-small cell lung cancer (NSCLC).
Catalog No. 50-208-4840 Supplier Creative Biomart Supplier No. THP0327
Hu-rhEGF-rP64k/Mont is a therapeutic vaccine composed by an antigen (recombinant human EGF (rhEGF) chemically conjugated to recombinant P64K (rP64K)) and an adjuvant (Montanide ISA51VG) being investigated for the treatment of advanced non-small cell lung cancer (NSCLC).