Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Activin RIB/ALK-4 protein, mouse (HEK293, Fc) | >80.0% | 10 UG
SDP

Catalog No. 5000291676
Encompass_Preferred

Recombinant mouse Activin RIB/ALK-4 extracellular domain produced in HEK293 cells with a C-terminal Fc tag. Supplied lyophilized for research applications requiring the receptor ectodomain and binding or signaling studies.

  • Size: 10 μg supplied lyophilized.
  • Species: mouse extracellular domain.
  • Expression system: HEK293 mammalian cells.
  • Tag: C-terminal Fc for detection and purification.
  • Amino acid range: L32-E126 (95 amino acids).
  • Molecular weight: 39-50 kDa.
  • Purity: >80% as determined by reducing SDS-PAGE.
  • Formulation: lyophilized from PBS, pH 7.4.
  • Storage: store at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.

Catalog No. 50-002-91676 Supplier Medchemexpress LLC Supplier No. HYP748610UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Activin RIB/ALK-4 extracellular domain produced in HEK293 cells with a C-terminal Fc tag. Supplied lyophilized for research applications requiring the receptor ectodomain and binding or signaling studies.

  • Size: 10 μg supplied lyophilized.
  • Species: mouse extracellular domain.
  • Expression system: HEK293 mammalian cells.
  • Tag: C-terminal Fc for detection and purification.
  • Amino acid range: L32-E126 (95 amino acids).
  • Molecular weight: 39-50 kDa.
  • Purity: >80% as determined by reducing SDS-PAGE.
  • Formulation: lyophilized from PBS, pH 7.4.
  • Storage: store at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.