Learn More
Genscript Corporation Activin RIIB/ACVR2BBiotin H
Catalog No. 5000059482
Encompass
ActRIIB (activin receptor type-2B) is an activin receptor subtype constitutively expressed in the whole body playing a role in cellular proliferation differentiation and metabolism For its various physiological activities ActRIIB interacts with activin and multiple other ligands including myostatin (MSTN) growth differentiation factor 11 (GDF11) and bone morphogenetic protein 9 (BMP9)
Catalog No. 50-000-59482 Supplier Genscript Corporation Supplier No. Z04391100
ActRIIB (activin receptor type-2B) is an activin receptor subtype constitutively expressed in the whole body playing a role in cellular proliferation differentiation and metabolism For its various physiological activities ActRIIB interacts with activin and multiple other ligands including myostatin (MSTN) growth differentiation factor 11 (GDF11) and bone morphogenetic protein 9 (BMP9)