Learn More
Activity Standard Solution, For Fluoride Electrode (100 ppm as fluoride) , MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Density | 1 g/mL (at 25°C) |
| Boiling Point | 100°C (750 mmHg) |
| Quantity | 475 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.