Learn More
Electron Microscopy Sciences Activated Carbon Filters, Manufacturer: Leica. Model: AutoStainer XL ST5010, MultiStainer ST5020, LinearStainer ST4040. Use: 3 months., 1/EA
SDP
Catalog No. 502090710
Encompass
The carbon filter cartridges contain a high grade activated carbon that effectively absorbs most organic and inorganic vapors, including, but not limited to, xylene, formalin, methanol, toluene, and isopropyl alcohol. The filters are an alternate to brand-name original carbon filters and perform the same, if not better, at a fraction of the cost. The sizing and fit is identical to brand name cartridges. Manufacturer: Leica. Model: AutoStainer XL ST5010, MultiStainer ST5020, LinearStainer ST4040 . Use: 3 months .
Catalog No. 50-209-0710 Supplier Electron Microscopy Sciences Supplier No. 6265401
The carbon filter cartridges contain a high grade activated carbon that effectively absorbs most organic and inorganic vapors, including, but not limited to, xylene, formalin, methanol, toluene, and isopropyl alcohol. The filters are an alternate to brand-name original carbon filters and perform the same, if not better, at a fraction of the cost. The sizing and fit is identical to brand name cartridges. Manufacturer: Leica. Model: AutoStainer XL ST5010, MultiStainer ST5020, LinearStainer ST4040 . Use: 3 months .