Learn More
Electron Microscopy Sciences Activated Carbon Filters, Manufacturer: Thermo Scientific Shandon™. Model: GLX Stainer, HyperClean Fume Hood, Consul/ClearVue Coverslipper, Excelsior Tissue Processor, Varistain Stainer/Gemini ES Stainer. Use: 3 months., 1/EA
SDP
Catalog No. 502090717
Encompass
The carbon filter cartridges contain a high grade activated carbon that effectively absorbs most organic and inorganic vapors, including, but not limited to, xylene, formalin, methanol, toluene, and isopropyl alcohol. The filters are an alternate to brand-name original carbon filters and perform the same, if not better, at a fraction of the cost. The sizing and fit is identical to brand name cartridges. Manufacturer: Thermo Scientific Shandon™. Model: GLX Stainer, HyperClean Fume Hood, Consul/ClearVue Coverslipper, Excelsior Tissue Processor, Varistain Stainer/Gemini ES Stainer . Use: 3 months .
Catalog No. 50-209-0717 Supplier Electron Microscopy Sciences Supplier No. 6265408
The carbon filter cartridges contain a high grade activated carbon that effectively absorbs most organic and inorganic vapors, including, but not limited to, xylene, formalin, methanol, toluene, and isopropyl alcohol. The filters are an alternate to brand-name original carbon filters and perform the same, if not better, at a fraction of the cost. The sizing and fit is identical to brand name cartridges. Manufacturer: Thermo Scientific Shandon™. Model: GLX Stainer, HyperClean Fume Hood, Consul/ClearVue Coverslipper, Excelsior Tissue Processor, Varistain Stainer/Gemini ES Stainer . Use: 3 months .