Learn More
Electron Microscopy Sciences Activated Carbon Filters, Manufacturer: Tissue-Tek Sakura. Model: DRS Stainer. Use: 3 months., 1/EA
SDP
Catalog No. 502090720
Encompass
The carbon filter cartridges contain a high grade activated carbon that effectively absorbs most organic and inorganic vapors, including, but not limited to, xylene, formalin, methanol, toluene, and isopropyl alcohol. The filters are an alternate to brand-name original carbon filters and perform the same, if not better, at a fraction of the cost. The sizing and fit is identical to brand name cartridges. Manufacturer: Tissue-Tek Sakura. Model: DRS Stainer . Use: 3 months .
Catalog No. 50-209-0720 Supplier Electron Microscopy Sciences Supplier No. 6265411
The carbon filter cartridges contain a high grade activated carbon that effectively absorbs most organic and inorganic vapors, including, but not limited to, xylene, formalin, methanol, toluene, and isopropyl alcohol. The filters are an alternate to brand-name original carbon filters and perform the same, if not better, at a fraction of the cost. The sizing and fit is identical to brand name cartridges. Manufacturer: Tissue-Tek Sakura. Model: DRS Stainer . Use: 3 months .