Learn More
Sentry Air Systems Model 400 Carbon Pre-filter Pads - 12 Pack
SDP
Supplier:  Sentry Air Systems SS400CFP
Encompass
Pack of 12 Carbon Pre-Filter Pads for Model 400 units. These pre-filter pad replacements help the filtration system to capture and remove harmful fumes and odors emitted from different processes. This pack fits all Model 400 fume extractors. The pre-filter is used in conjunction with a Model 400 HEPA, ASHRAE, ULPA, Activated Carbon, or specialty-blended filter.
Catalog No. NC1133268