Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ActivX™ Azido-FP Serine Hydrolase Probe

Catalog No. PI88316 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
3.5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88316 3.5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88316 Supplier Thermo Scientific™ Supplier No. 88316
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ActivX™ probes have been designed to selectively label and enrich serine hydrolases through tagged fluorophosphonate to assess enzyme activity. ActivX Serine Hydrolase Probes covalently label serine hydrolase using fluorophosphonate conjugated with either desthiobiotin or azide.

ActivX™ FP Probes feature a reactive fluorophosphonate group that specifically and covalently labels the active-site serine of enzymatically active serine hydrolases. These probes are available with a desthiobiotin (biotin analog) tag for selective enrichment or a reactive- azido group (Staudinger reagents) that facilitate multiplex labeling when used with phosphine- or alkyne-derivatized tags.

ActivX active-site enrichment probes are advantageous for determining active enzyme levels using analog-tagged probes to allow profiling of both inactive and active enzymes. The ActivX probes selectively enrich only those enzymes that are functionally active and biologically relevant at the time of labeling. With selective enrichment, one can identify and profile target enzyme classes across samples or can assess the specificity and affinity of enzyme inhibitors.

Applications of ActivX FP Probes:

  • Determine serine hydrolase enzyme activity in cells and lysates
  • Mapping the active-site serine of functionally diverse serine hydrolase family members (e.g. proteases, lipases, esterases)
  • Screen for small molecule binding affinities and active-site inhibition
  • Profile serine hydrolases using fluorescent, Western blot or mass spectrometry workflows

The serine hydrolase superfamily is one of the largest, most diverse enzyme families in eukaryotic proteomes. Serine hydrolases are generally grouped into two large families: serine proteases (e.g., trypsin, elastase and thrombin) and metabolic serine hydrolases. Metabolic serine hydrolases are divided into multiple enzyme subclasses (e.g., esterases, lipases, amidases and peptidases) based on structure, catalytic mechanism and substrate preference. These probes can be used to assess activity or screen small molecule inhibitors against enzymes derived from cell lysates, subcellular fractions, tissues and recombinant proteins.

Assessment of active-site labeling with desthiobioin probes can be accomplished by either Western blot or mass spectrometry (MS). For the Western blot workflow, desthiobiotin-labeled proteins are enriched for SDS-PAGE analysis and subsequent detection with specific antibodies. For the MS workflow, desthiobiotin-labeled proteins are reduced, alkylated and enzymatically digested to peptides. Only the desthiobiotin-labeled, active-site peptides are enriched for analysis by LC-MS/MS. Both workflows can be used for determining inhibitor target binding, but only the MS workflow can identify global inhibitor targets and off-targets. The Azido-FP probe can be used in combination with a phosphine- or alkyne-derivatized tag for either detection or enrichment of serine hydrolases using Western blot or mass spectrometry.

These products are subject to a limited use label license.

TRUSTED_SUSTAINABILITY

Specifications

Description ActivX™ Azido-FP Serine Hydrolase Probe
Formulation Azido-fluorophosphonate, MW 350
Content And Storage Store at -80°C away from light. Stock solutions in DMSO can be stored at -20°C for 6 months.
Assay Protein Quantitation Assay
Detection Method Chemiluminescence, Mass Spectrometry, Western Blot
Substrate Type Serine Hydrolase Substrate
Product Line ActivX
Quantity 3.5 μg
For Use With (Application) Pull-Down
Target Serine Hydrolase
Type Fluorophosphonate (FP) probe
Label or Dye Click Chemistry Label
Format Tube, Spin Column
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.