Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ActivX™ TAMRA-FP Serine Hydrolase Probe

Catalog No. PI88318 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
6.8 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI88318 6.8 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI88318 Supplier Thermo Scientific™ Supplier No. 88318
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

ActivX™ probes have been designed to selectively label and enrich serine hydrolases through binding of their respective active sites with tagged fluorophosphonate to enable assessment of enzyme activity.

The TAMRA-FP probe feature a reactive fluorophosphonate group that is used to label and detect active serine hydrolases by fluorescent gel imaging, capillary electrophoresis or mass spectrometry. An anti-TAMRA antibody is also available for immuno-enrichment of TAMRA-FP probe-labeled proteins.

ActivX™ active-site enrichment probes are advantageous for determining active enzyme levels through the use of analog-tagged probes to allow profiling of both inactive and active enzymes. The ActivX probes selectively enrich only those enzymes that are functionally active and biologically relevant at the time of labeling. With selective enrichment, one can identify and profile target enzyme classes across samples or can assess the specificity and affinity of enzyme inhibitors.

The serine hydrolase superfamily is one of the largest, most diverse enzyme families in eukaryotic proteomes. Serine hydrolases are generally grouped into two large families: serine proteases (e.g., trypsin, elastase and thrombin) and metabolic serine hydrolases. Metabolic serine hydrolases are divided into multiple enzyme subclasses (e.g., esterases, lipases, amidases and peptidases) based on structure, catalytic mechanism and substrate preference. These probes can be used to assess activity or screen small molecule inhibitors against enzymes derived from cell lysates, subcellular fractions, tissues and recombinant proteins.

These products are subject to a limited use label license.

TRUSTED_SUSTAINABILITY

Specifications

Description ActivX™ TAMRA-FP Serine Hydrolase Probe
Formulation TAMRA-fluorophosphonate, MW 680
Content And Storage Store at -80°C away from light. Stock solutions in DMSO can be stored at -20°C for 6 months.
Assay Protein Quantitation Assay
Detection Method Chemiluminescence, Mass Spectrometry, Western Blot
Substrate Type Serine Hydrolase Substrate
Product Line ActivX
Quantity 6.8 μg
For Use With (Application) Pull-Down
Target Serine Hydrolase
Type Fluorophosphonate (FP) probe
Label or Dye Other Label(s) or Dye(s)
Format Tube, Spin Column
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.