Learn More
Cayman Chemical Acetyl Dipeptide 1 cetyl ester (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502910657
Encompass_Preferred
A cosmeceutical peptide; increases expression of the genes encoding the skin barrier function proteins AQP3, FLG, caspase-14, and cytokeratin 10 in primary human keratinocytes; reduces glyoxal-induced AGE formation in primary human fibroblasts
Catalog No. 50-291-0657 Supplier Cayman Chemical Supplier No. 3494910 MG
A cosmeceutical peptide; increases expression of the genes encoding the skin barrier function proteins AQP3, FLG, caspase-14, and cytokeratin 10 in primary human keratinocytes; reduces glyoxal-induced AGE formation in primary human fibroblasts