Learn More
Aviva Systems Biology Corp ACTL6B Antibody - middle region (ARP56847_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56847_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACTL6B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ACTL6B. Alternate names: ACTL6, BAF53BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62010