Learn More
Aviva Systems Biology Corp ACTN2 Antibody - C-terminal region (ARP41463_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41463_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ACTN2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human ACTN2. Alternate names: CMD1AAApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53301