Learn More
Aviva Systems Biology Corp ACTN4 Antibody - N-terminal region (ARP42202_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42202_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ACTN4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACTN4. Alternate names: DKFZp686K23158, FSGS, FSGS1, ACTININ-4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58139