Learn More
Aviva Systems Biology Corp ACTN4 Antibody - C-terminal region (ARP42203_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42203_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACTN4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ACTN4. Alternate names: DKFZp686K23158, FSGS, FSGS1, ACTININ-4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-58140