Learn More
Cayman Chemical Acetoacetyl-Coenzyme A (sodium salt hydrate), 10 mg
SDP
Catalog No. 502981144
Encompass_Preferred
A precursor to HMG-CoA in the isoprenoid pathway; reversibly converted to acetyl-CoA by acetoacetyl-CoA thiolase
Catalog No. 50-298-1144 Supplier Cayman Chemical Supplier No. 2121910 MG
A precursor to HMG-CoA in the isoprenoid pathway; reversibly converted to acetyl-CoA by acetoacetyl-CoA thiolase